Learn More
Medchemexpress LLC PROTAC TYK2 degradation agent1 | 2921556-14-9 | C55H69N13O7S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY152227
Encompass_Preferred
PROTAC TYK2 degradation agent1 is a potent and subtype-selective TYK2 degrader with a DC50 value of 14 nM for TYK2 degradation activity. It is intended for research use, particularly in the study of autoimmune diseases.
  • Potent and subtype-selective TYK2 degrader
  • DC50 value of 14 nM for TYK2 degradation activity
  • Selectively suppresses TYK2 protein levels
  • Effectively induces degradation of TYK2 in a dose-dependent manner at low concentrations
  • Can be used for research of autoimmune diseases
Catalog No. 50-003-75403