Learn More
Selleck Chemical LLC Protamine sulfate P1169-100MG
SDP
Supplier:  Selleck Chemical LLC P1169100MG
Encompass_Preferred
Protamine sulfate is a polycationic highly positively charged protein derived from salmon sperm protein used to neutralize unfractionated heparin administration (UFH)-induced anticoagulation following cardiovascular surgical procedures
Catalog No. 50-000-40846