Learn More
Enzo Life Sciences Proteasome 19S Rpt1/S7 subunit (human) monoclonal antibody (MSS1-92) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW94000025
Encompass_Preferred
Clone: MSS1-92. Isotype: IgG1. Immunogen: Recombinant human Rpt1 protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: Recognizes the Rpt1/S7 subunit of the 19S regulator complex. Applications: IP. Purity: Ammonium sulphate precipitation and ion-exchanging chromatography purified. Formulation: Liquid. In TRIS buffer containing 10mM sodium azide.
Catalog No. 50-201-1686