Learn More
AG Scientific Inc Protease Inhib Cocktail I
Supplier:  AG Scientific Inc P1510VIAL
Encompass_Preferred
"Protease Inhibitor Cocktail I is a mixture of five protease inhibitors which has been optimized for the inhibition of a broad range of proteases and esterases. 1x stock solution contains: 500 μM AEBSF HCl 150 nM Aprotinin 1 μM E-64 0.5 mM EDTA disodium salt 1μM Leupeptin Hemisulfate. Preparation: Reconstitute each vial with 1 mL water to obtain a 100X concentrated stock solution."
Catalog No. 50-252-2615