Learn More
AG Scientific Inc Protease Inhib VI Plant Cells
Supplier:  AG Scientific Inc P1545VIAL
Encompass_Preferred
"Protease Inhibitor Cocktail VI is a specially formulated cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, serine, metalloproteases as well as aminopeptidases. Recommended for use with plant cell extracts. Supplied as liquid in 1 mL DMSO, each vial contains: 200 mM AEBSF HCl 10 mM bestatin 3 mM E-64 2 mM Leupeptin hemisulfate 500 mM o-phenanthroline 2 mM Pepstatin A Preparation: Add to water, aqueous buffer or directly to extraction media. 1 vial is recommended for about 30 g of plant tissues. "
Catalog No. 50-252-2603