Learn More
Apexbio Technology LLC Protease Inhibitor Cocktail (EDTA-Free, 200X in DMSO), 1ml x 50
SDP
Supplier:  Apexbio Technology LLC K100850
Encompass_Preferred
Equivalent to Sigma P1860, for use in tissue culture media to prevent proteolytic degradation of secreted proteins. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5537