Learn More
Bioworld Protease Inhibitor Cocktail I Animal Free for General Use, 1 Vial(s)
Supplier:  Bioworld 220200031
Encompass_Preferred
Lyophillzed blend of five protease inhibitors for applications that require animal free reagents. Reconstitute with 1 ml water to produce a 100X concentrated solution.* Contains: AEBSF Aprotinin (recombinant) E-64 EDTA Leupeptin* Specificity: Serine Proteases, Esterases, Cysteine Proteases, Metalloproteases and Trypsin like ProteasesThis product is sold for laboratory use only.
Catalog No. 50-252-984