Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp PROTEASE INHIBITOR COCKTAIL I,ANIMAL FREE,FOR GENERAL USE, 1 VIAL
SDP

Supplier:  Research Products International Corp P506501

Encompass_Preferred

Lyophillzed blend of five protease inhibitors for applications that require animal free reagents. Reconstitute with 1 ml water to produce a 100X concentrated solution.

Contains:

AEBSF
Aprotinin (recombinant)
E-64
EDTA
Leupeptin

Specificity:
Serine Proteases
Esterases
Cysteine
Proteases
Metalloproteases and Trypsin like Proteases

Catalog No. 50-488-955


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.