Learn More
Bioworld Protease Inhibitor Cocktail I, 5 mL
Supplier:  Bioworld 220200022
Encompass_Preferred
Protease Inhibitor Cocktail I (10x) SolutionAEBSF E-64 Aprotinin EDTA Leupeptin Store -20°C
Catalog No. 50-252-983