Learn More
Research Products International Corp Protease Inhibitor Cocktail II, for Bacterial Cell Extracts, Dry Powder, 1 Vial

Supplier: Research Products International Corp P508001
Lyophilized mixture of five protease inhibitors for use with bacterial cell extracts. Reconstitute with 1ml DMSO and 4ml water to obtain a 5ml solution. Will treat proteases extracted from 20g E.Coli.
Store at -20°:C
Contains: AEBSF, EDTA, Bestatin, E-64, Pepstatin A
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.