Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Protease Inhibitor Cocktail V, Animal Free, EDTA Free, 1 Vial
SDP

Supplier:  Research Products International Corp P509501

Encompass_Preferred

Lyophillized blend of four protease inhibitors for use in applications that require animal free reagents. Reconstitute with lml water to produce a 100X concentrated solution.

Contains:

AEBSF
Aprotinin (recombinant)
E-64
Leupeptin

Specificity:
Serine Proteases
Cysteine Proteases
Trypsin like Proteases

Catalog No. 50-136-8533


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.