Learn More
Selleck Chemical LLC Protease Inhibitor Library L2500-2X100UL/WELL
SDP
Supplier:  Selleck Chemical LLC L25002X100UL/WELL
Encompass_Preferred
A unique collection of 454 small molecule inhibitors used for chemical genomics high-throughput screening (HTS) and high content screening (HCS)
Catalog No. 50-000-40834
Add to Cart
Request a Quote