Learn More
Supply Solutions Protease Inhibitor Cocktail, MFCD00677817, 1mL
Supplier:  Supply Solutions SIGMA P83401ML
Encompass_Preferred
For use with mammalian cell and tissue extracts, DMSO solution, Inhibits serine, cysteine, and acid proteases, and aminopeptidases.
Catalog No. NC0829187