Learn More
Abcam Protease Inhibitor Cocktail EDTA Free (100 x ), 3ML
Supplier:  Abcam AB2700553ML
Encompass_Preferred
Protease inhibitor cocktail EDTA free (100X) protects protein extracts from aminopeptidases, metalloproteases, and serine, cysteine, and aspartic acid proteases. - Suitable for protease inhibition during protein extraction from mammalian tissues and cells, yeast and fungal cells, and bacteria - 100X stock solution of protease inhibitors dissolved in DMSO - Does not contain the chelating agent EDTA
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-196-3753