Learn More
Medchemexpress LLC HY-108717 100mg Medchemexpress, Protease K CAS:39450-01-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108717100MG
Encompass_Preferred
Medchemexpress, HY-108717 100mg Protease K CAS:39450-01-6 Protease K is a nonspecific serine protease that is useful for general digestion of proteins. Protease K is active in the presence of SDS or urea and over a wide range of pH (4-12), salt concentrations, and temperatures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0753