Learn More
Enzo Life Sciences Proteasome 20S beta5 subunit (human) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW88950100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 194-207 of human proteasome 20S (β5 subunit). Source: Purified from rabbit serum. Species reactivity: Human. Specificity: Recognizes human proteasome 20S β5 subunit. Applications: IHC, WB. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-1655