Learn More
Medchemexpress LLC Proteasome 20S C2/HC2 antibody (YA2301) | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8255650UL
Encompass_Preferred
Proteasome 20S C2/HC2 Antibody (YA2301) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to Proteasome 20S C2/HC2. It is for research use only and not intended for sale to patients. This antibody is a component of the 20S core proteasome complex involved in the proteolytic degradation of most intracellular proteins. It plays numerous essential roles within the cell by associating with different regulatory particles, participating in ATP-dependent degradation of ubiquitinated proteins and ubiquitin-independent protein degradation.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets Proteasome 20S C2/HC2
  • For research use only
  • Involved in proteolytic degradation of most intracellular proteins
  • Essential for maintaining protein homeostasis
  • Mediates both ATP-dependent and ubiquitin-independent protein degradation
Catalog No. 50-004-43296