Learn More
Medchemexpress LLC Proteasome 20S LMP7 Antibody (YA684) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8087610UL
Encompass_Preferred
Proteasome 20S LMP7 Antibody (YA684) is a mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting Proteasome 20S LMP7. This antibody exhibits reactivity with human and rat samples and is utilized in cell biology research.
  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets Proteasome 20S LMP7
  • Reactivity with human and rat samples
  • Predicted and observed band size of 30 kDa
  • Affinity purified
  • Applications include Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunohistochemistry-Frozen (IHC-F), and Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Essential for processing class I MHC peptides
Catalog No. 50-004-42611