Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Proteasome inhibitor IX | 856849-35-9 | 99.4% | 377.01 g·mol⁻1 | C20H21B2NO5 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10741210MM 1ML

Encompass_Preferred

Proteasome inhibitor IX is a chalcone-derived small molecule inhibitor of the 20S proteasome that preferentially inhibits chymotrypsin-like activity (reported IC50 ≈ 1 μM) and shows antiproliferative activity in cell-based assays.

  • Inhibits chymotrypsin-like activity of the 20S proteasome (IC50 ≈ 1 μM).
  • Demonstrates antiproliferative activity in HCT116 p53+/+ cells (IC50 ≈ 1.49 μM).
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • High purity, 99.4% (HPLC).
  • Molecular weight 377.01 g·mol⁻1.
  • Molecular formula C20H21B2NO5.

Catalog No. 50-002-40723


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.