Learn More
Essendant PROTECSORCVRLXLHDWH25
Supplier:  Essendant ANS46114
Encompass_Preferred
Features Patented Reflex design which offers excellent fit (with elastic back) and freedom of movement The material is strong and abrasion resistant and filters out many water-based liquids and dry particles Hooded Elastic Wrist and Ankles 1-Inch Flap Apparel Type- Coverall Material(s)- MICROFORCE Barrier SMS Fabric Color(s)- White Color Family- White
Catalog No. 50-000-22918