Learn More
Cayman Chemical ProtectIn D1 methyl ester 10ug
SDP
Supplier:  Cayman Chemical 3674610 UG
Encompass_Preferred
An esterified form of protectin D1; an intermediate in the synthesis of protectin D1; reduces infarct area and volume and increases blood vessel density in the tissue surrounding the infarct in a rat model of MCAO-induced ischemic stroke
Catalog No. 50-291-3262