Learn More
JDM LAB SOLUTIONS LLC Protective cover for SmartRack
Supplier:  JDM LAB SOLUTIONS LLC 01608
Encompass
The protective hood made of fiber-reinforced polyethylene (mesh film), which was specially designed for the SmartRack , reliably protects the SmartRack against dirt and dust particles as well as providing splash protection against aggressive liquids. The dust and particle protection hood is available in two versions that do not differ in size. As a basic model and as a model with two zippers on the edges of the front. For regular removal of the dust and particle protection hood and for use in unfavorable height conditions, we recommend choosing the model with zippers.
Catalog No. 50-255-8519