Learn More
Grainger Protective Leg Tip SlipOn Rd 13/32 PK10
Supplier:  Grainger 16ZF82
Encompass_Preferred
The Grainger 16ZF82 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face from sparks, spatter, and harmful UV/IR rays while welding. This helmet has a large viewing area for increased visibility. It has adjustable sensitivity and delay controls so you can customize settings for each application. The helmet operates on solar cells with lithium backup batteries for convenience. The Speedglas design provides exceptional comfort and balance. This auto-darkening welding helmet meets ANSI Z87.1 safety standards. With its advanced optics and rugged durability, this helmet is ideal for professional welders.
Catalog No. 50-276-8569