Learn More
Grainger Protective Leg Tip SlipOn Rd 19/32 PK25
Supplier:  Grainger 16ZE11
Encompass_Preferred
The Grainger 16ZE11 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It has Class F insulation and is UL/CSA certified. The 16ZE11 operates on 115V power and has a full load amp rating of 12.2A.
Catalog No. 50-276-5456