Learn More
Grainger Protective Leg Tip SlipOn Rd 3/4 PK25
Supplier:  Grainger 16ZE16
Encompass_Preferred
The Grainger 16ZE16 is a 16 oz. can of 3M Super 77 Multipurpose Spray Adhesive. It is a high strength, fast-tacking spray adhesive that bonds lightweight materials like paper, cardboard, foam, felt, fabrics, and more. The web-spray pattern provides efficient coverage. It has high initial tack and fast drying. The can contains no CFCs or other ozone depleting substances. It is made in the USA. This versatile spray adhesive is useful for crafts, office use, home DIY projects, and more. The 16 oz can provides extended usage.
Catalog No. 50-276-6329