Learn More
Grainger Protective Leg Tip SlipOn Rd 39/64 PK25
Supplier:  Grainger 16ZM82
Encompass_Preferred
The Grainger 16ZM82 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This UL rated, 2.5 lb extinguisher uses Novec 1230 fluid which leaves no residue and is ideal for protecting valuable equipment and electronics. The extinguisher has a discharge range of 10-12 ft and can operate between -4°F to 120°F. It features an all-steel valve construction and comes with a wall hook for easy mounting. The extinguisher is compact at only 5.4"W x 16.5"H and is sold fully charged.
Catalog No. 50-276-8568