Learn More
Grainger Protective Leg Tip SlipOn Rd 47/64 PK25
Supplier:  Grainger 16ZM91
Encompass_Preferred
The Protective Leg Tip Slip-On by Protective Industrial Products is a leg tip protector designed to prevent damage to furniture legs and floors. It is made of clear vinyl plastic and has a ribbed surface for grip. The leg tip cover measures 2" in diameter and slides onto table and chair legs up to 1 3/4" in diameter. It is sold in a pack of 25 leg tips. The leg tip covers provide a cushion between furniture legs and floors to prevent scratches, scuffs and noise. They are easy to install by sliding onto the bottom of furniture legs.
Catalog No. 50-276-8564