Learn More
Grainger Protective Leg Tip SlipOn Rd 63/64 PK25
Supplier:  Grainger 16ZF55
Encompass_Preferred
The Protective Leg Tip Slip-On RD-63-64 by Protective Industrial Products is a slip-on leg tip for round tubing with an outside diameter of 0.625 inches. It is made of molded PVC and helps protect floors and equipment from damage caused by metal tubing ends. The leg tips are designed for use on materials handling equipment, shelving, racks, carts, and furniture. They simply slip over the end of the tubing for quick installation. The leg tips are black in color and sold in packs of 25.
Catalog No. 50-276-5778