Learn More
Grainger Protective Sleeve 25 L 2.16 W Black
Supplier:  Grainger 16P896
Encompass_Preferred
The Grainger Protective Sleeve is a 25 foot long, 2 inch wide black polyester sleeve designed to protect cables and hoses from abrasion. It features a zipper closure for easy installation and removal and has a temperature rating from -20°F to 160°F. The sleeve's polyester construction makes it durable and flexible. With its 25 foot length, it can cover long cable and hose runs. The 2 inch width allows it to accommodate cables and hoses up to 1.75 inches in diameter. This abrasion-resistant sleeve helps prolong the life of cables and hoses in industrial environments.
Catalog No. 50-276-3927