Learn More
Essendant PROTECTOREARMUFFGN
Supplier:  Essendant MMMX1A
Encompass_Preferred
Modern and attractive PELTORxSeries Earmuffs provide excellent hearing protection against a wide range of noise sources arising within both workplace and leisure-activity environments Low-profile lightweight design is perfectly balanced to deliver superior wearing comfort Padded easy-adjust twin-wire headband is fully encased within a non-conductive material to protect against low-voltage electrical hazards (less than 440 V AC) Pivot/tilt earcups and soft wide cushions help the wearer achieve and maintain the proper amount of pressure around the ears to ensure an excellent highly effective acoustic seal without compromising the earmuff s snug pleasant fit Innovative foam earcup inserts and spacers further improve noise-attenuation performance Cushions and inserts are removable and easy-to-replace (replacement cushions and inserts sold separately) Model X1A For low to moderate noise-exposure environments Material(s)- ABS Plastic Polyester Polyvinyl Chloride (PVC) Stainl
Catalog No. 50-000-35970