Learn More
Bioworld PROTEIN-A BLUE SEPAROPORE
Supplier:  Bioworld 201810421
Encompass
This item has a minimum order quantity of 2 per supplier requirements. PROTEIN-A BLUE SEPAROPORE (AGAROSE) 4B (MACROBEADS): Protein A Blue Separopore 4B (Macrobeads) Affinity matrix with immobilized Protein-A by CNBr activation coupling method.; For purification and/or fractionation of IgG subclasses.; Rel...
Catalog No. 50-197-770