Learn More
Cygnus Technologies PROTEIN A ELISA KIT
Supplier:  Cygnus Technologies F400Z
Encompass
This NEWPROTEINAELISA kit is designed to overcome low analytical recovery of Protein A encountered in many commercial kits. Most low recovery samples are either incomplete antibody fragments or so-called fusion proteins.
Catalog No. NC0430610