Learn More
Cygnus Technologies PROTEIN A ELISA KIT
Supplier:  Cygnus Technologies F400
Encompass
This ELISA incorporates some improvements over the earlier kits and is designed to better detect the newer, unnatural constructs of Protein A such as the MabSelect SuReTM ligand sold by GE Healthcare. If you are using a recombinant form of Protein A with very significant structural differences from
Catalog No. NC9632097