Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein A SepharoseR from5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P33911.5G
Encompass
Protein A-Sepharose when linked with antibody helps improving antigen binding capacity of IgG crosslinked to column matrices. In this case binding happens between Protein A and fc portion of the IgG molecule leaving antigen specific sites free.
Catalog No. 50-291-4224