Learn More
Abcam Protein Block, 125ML
Supplier:  Abcam AB64226125ML
Encompass_Preferred
Protein block is a serum-free blocking buffer designed to reduce non-specific background staining in IHC, ELISA, and western blot (WB). - Cited in over 90 publications - No need to match species for secondary antibodies, reducing animal serum handling - Ready-to-use – no mixing or dilution required
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0779158