Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein Carbonyl Content A
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK0941KT
Encompass
Oxidative stress results when the effectiveness of antioxidant defenses is insufficient to deal with the production of reactive oxygen species (ROS). ROS can induce damage to DNA lipids and proteins. The oxidation of proteins results in the production of stable carbonyl groups which can be used as a measure of oxidative injury.
Catalog No. 50-290-4144