Learn More
Sino Biological Protein Cyno IL13 ET-FREE 100
Supplier:  Sino Biological 11057C07HUE100
Encompass_Preferred
A DNA sequence encoding the mature form of Cynomolgus (Macaca fascicularis) IL-13 (ABG75889 1) (Ser 21-Asn 132) was fused with a polyhistidine tag at the N-terminus
Catalog No. 50-010-73836