Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protein G Agarose | 50 ML
SDP

Supplier:  Medchemexpress LLC HYK021450ML

Encompass_Preferred

Protein G is a bacterial cell wall protein isolated from group G Streptococci that binds to most mammalian IgGs primarily through Fc regions. Recombinant Protein G, with its albumin and cell surface binding sites eliminated to reduce nonspecific binding, is coupled with 4% highly cross-linked agarose in this product. It effectively purifies mammalian monoclonal and polyclonal antibodies, including human IgG3 and rat IgG2a.

  • Composition: 4% cross-linked agarose
  • Degree of substitution: 2 mg recombinant protein G/mL settled resin
  • Bead diameter: 45-165 µm
  • Binding capacity: 20 mg human IgG/mL settled resin
  • Storage: 50% slurry in 1× PBS with 20% ethanol; stable for at least 2 years at 2-8°C; do not dry or freeze.

Catalog No. 50-004-30572


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.