Learn More
Sino Biological Recombinant Human AKT2 Protein (Full length, TST & Trx & TEV Tag) 10
Supplier:  Sino Biological A1710T10
Encompass_Preferred
Recombinant full-length human AKT2 was expressed by baculovirus in Sf9 insect cells using an N-terminal TST-Trx-TEV tag.
Catalog No. 50-006-99123