Learn More
Sino Biological Recombinant Human ATAD2 Protein (Bromodomain, His Tag), Active 500 µg
Supplier:  Sino Biological A22310H500
Encompass_Preferred
Recombinant human ATAD2A bromodomain (981-1108) was expressed in E. coli cells using an N-terminal His tag.
Catalog No. 50-006-56654