Learn More
Sino Biological Recombinant Human CTLA-4 Protein (AF 647 site-specific conjugation, His Tag) 25
Supplier:  Sino Biological 11159H86HSG25
Encompass_Preferred
A DNA sequence encoding the human CTLA4 (NP005205.2) (Lys36-Phe162) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with AF 647 (Excitation Max.= 655 nm, Emission Max.= 680 nm).
Catalog No. 50-006-99439