Learn More
Sino Biological Recombinant Human Fc epsilon RI Protein (hFc & AVI Tag), HPLC-verified 20
Supplier:  Sino Biological 13193H41HB20
Encompass_Preferred
A DNA sequence encoding the Human FCER1A (P12319)(Met1-Gln205) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-99402