Learn More
Sino Biological Protein Human GHR ET FREE1MG
Supplier:  Sino Biological 17302H08HUE1
Encompass_Preferred
A DNA sequence encoding the Human GHR (P10912) (Ala27-Tyr264) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-99009