Learn More
Sino Biological Recombinant Human Interferon alpha 4/IFNA4 Protein (His Tag) 20 µg
Supplier:  Sino Biological 10336H08B20
Encompass_Preferred
A DNA sequence encoding the human IFNα 4a (NP_066546.1)) (Met 1-Asp 189) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-99450