Learn More
Sino Biological Recombinant Human IL-31RA Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified 20
Supplier:  Sino Biological 15878H49HB20
Encompass_Preferred
A DNA sequence encoding the Huamn IL-31RA (EAW54932.1) (Met1-Thr516) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-99444