Learn More
Sino Biological Recombinant Human NCAM-1/CD56 Protein (Site-Specific AF 647 Conjugation, His Tag) 100 µg
Supplier:  Sino Biological 10673H86HSG100
Encompass_Preferred
A DNA sequence encoding the human NCAM1 (NP_001070150.1) (Met 1-Pro 603) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with AF 647 (Excitation Max.= 655 nm, Emission Max.= 680 nm).
Catalog No. 50-006-56813