Learn More
Sino Biological Recombinant Human NCAM-1/CD56 Protein (Site-Specific APC Conjugation, His Tag) 25 Tests
Supplier:  Sino Biological 10673H86HSA25
Encompass_Preferred
A DNA sequence encoding the human NCAM1 (NP_001070150.1) (Met 1-Pro 603) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with APC (Excitation Max.= 651 nm, Emission Max.= 662 nm).
Catalog No. 50-006-56643