Learn More
Sino Biological Protein Human NCR1 100UG
Supplier:  Sino Biological 30142H08H1UE100
Encompass_Preferred
A DNA sequence encoding the Human NCR1 (O76036-1) (Gln22-Gln254) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-99101