Learn More
Sino Biological Recombinant Human NR1H3/LXR alpha Protein (Full length, TST-GST-TEV Tag) 100 µg
Supplier:  Sino Biological L3130T100
Encompass_Preferred
Recombinant full-length human LXR alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal TST-GST-TEV tag.
Catalog No. 50-006-56591