Learn More
Sino Biological Protein Human PODXL 100UG
Supplier:  Sino Biological 29798H02HUE100
Encompass_Preferred
A DNA sequence encoding the Human PODXL (O00592) (Ser23-Pro461) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-006-98867